Synonyms:
Bis(cyclopentadienyl)zirconium(IV) chloride hydride, Schwartz's Reagent, Bis(cyclopentadienyl)zirconium chloride hydride
L15164.06
L15164.06EA
138
EUR
InStock
L15164.06
Bis(cyclopentadienyl)zirconium hydridochloride 95%
Bis(cyclopentadienyl)zirconium hydridochloride
Purity: 95%
CAS No.: 37342-97-5
Molecular Formula: C10H11ClZr
Formula Weight: 257.87
Storage Temperature: 2°C to 8°C
Melting Point: 300°
UN No.: 3131
MDL No.: MFCD02089401