L15252.06
L15252.06EA
87.4
EUR
InStock
L15252.06
Isocyanate de 2-éthylphényle ≥99%
Isocyanate de 2-éthylphényle
Purity: 99%
CAS No.: 40411-25-4
Molecular Formula: C9H9NO
Formula Weight: 147.18
Storage Temperature: 2°C to 8°C
Flash Point: 78°(172°F)
Refractive Index: 1.5280
Boiling Point: 56°/3mm
Beilstein Reference: 3538910
UN No.: 2206
MDL No.: MFCD00002013