L15993.06
L15993.06EA
91.8
EUR
InStock
L15993.06
Isocyanate de 3-chloropropyle ≥97%
Isocyanate de 3-chloropropyle
Purity: 97%
CAS No.: 13010-19-0
Molecular Formula: Cl(CH2)3NCO
Formula Weight: 119.55
Storage Temperature: 2°C to 8°C
Flash Point: 61°(141°F)
Refractive Index: 1.4520
Odour: Pungent
Boiling Point: 152-154°
Beilstein Reference: 1098789
UN No.: 2206
MDL No.: MFCD00010384