Stabilization:
Stabilised with hydroquinone (HQ)
L16274.18
L16274.18EA
33.4
EUR
InStock
L16274.18
2,4,6-Triallyloxy-1,3,5-triazine ≥98% stabilisé
2,4,6-Triallyloxy-1,3,5-triazine
Stabilized with 100 ppm hydroquinone
Purity: 98%
CAS No.: 101-37-1
Molecular Formula: C12H15N3O3
Formula Weight: 249.27
Flash Point: 166°(331°F)
Melting Point: 25-28°
Boiling Point: 149-150°/4mm
UN No.: 3077
MDL No.: MFCD00006049