Synonyms:
2-Hydroxy-5-(trifluoromethoxy)benzaldehyde
L17136.03
L17136.03EA
144
EUR
InStock
L17136.03
5-(Trifluoromethoxy)salicylaldehyde ≥98%
5-(Trifluoromethoxy)salicylaldehyde
Purity: 98+%
CAS No.: 93249-62-8
Molecular Formula: C8H5F3O3
Formula Weight: 206.12
Flash Point: 110°(230°F)
Melting Point: 31-33°
Boiling Point: 82°/60mm
Beilstein Reference: 7023825
MDL No.: MFCD00075249