L17285.06
L17285.06EA
89.5
EUR
InStock
L17285.06
Methoxyflurane 97%
Methoxyflurane
Purity: 97%
CAS No.: 76-38-0
Molecular Formula: CH3OCF2CHCl2
Formula Weight: 164.97
Flash Point: 37°(98°F)
Melting Point: -36°
Boiling Point: 105°
Beilstein Reference: 1737766
UN No.: UN3271
MDL No.: MFCD00040144