L17440.06
L17440.06EA
106
EUR
InStock
L17440.06
1-Bromo-3-(tert-butyldimethylsiloxy)benzene ≥98%
1-Bromo-3-(tert-butyldimethylsiloxy)benzene
Purity: 98+%
CAS No.: 65423-56-5
Molecular Formula: C12H19BrOSi
Formula Weight: 287.27
Refractive Index: 1.5090
Boiling Point: 66°/0.3mm
Beilstein Reference: 2256298
MDL No.: MFCD01318106