L17549.06
L17549.06EA
56.2
EUR
InStock
L17549.06
N-(4-Carboxyphenyl)succinamic acid 99%
N-(4-Carboxyphenyl)succinamic acid
Purity: 99%
CAS No.: 76475-62-2
Molecular Formula: C11H11NO5
Formula Weight: 237.21
Melting Point: ca 290° dec.
Beilstein Reference: 3355574
MDL No.: MFCD00029834