L17671.MD
L17671.MDEA
146
EUR
InStock
L17671.MD
DL-2-Amino-4-phosphonobutyric acid 95%
DL-2-Amino-4-phosphonobutyric acid
Purity: 95%
CAS No.: 20263-07-4
Molecular Formula: C4H10NO5P
Formula Weight: 183.10
Melting Point: 225-228°
Beilstein Reference: 2093908
MDL No.: MFCD00013999