L18428.06
L18428.06EA
108
EUR
InStock
L18428.06
4-sulphamidobenzoyl chloride DMF complex 95%
4-sulphamidobenzoyl chloride DMF complex
Purity: 95%
CAS No.: 1172493-99-0
Molecular Formula: C10H13ClN2O4S
Formula Weight: 292.74
UN No.: UN3261
MDL No.: MFCD02094019