L18432.03
L18432.03EA
47.1
EUR
InStock
L18432.03
4-Acetamidobenzoyl chloride 95%
4-Acetamidobenzoyl chloride
Purity: 95%
CAS No.: 16331-48-9
Molecular Formula: C9H8ClNO2
Formula Weight: 197.62
Melting Point: ca 145° dec.
UN No.: UN3261
MDL No.: MFCD02094022