Synonyms:
(5-Cyanothiophen-2-yl)boronic acid, 5-Cyanothiophene-2-boronic acid
L18523.03
L18523.03EA
67.3
EUR
InStock
L18523.03
5-Cyano-2-thiopheneboronic acid ≥98%
5-Cyano-2-thiopheneboronic acid
Purity: 98%
CAS No.: 305832-67-1
Molecular Formula: C5H4BNO2S
Formula Weight: 152.97
Melting Point: 125-128°
MDL No.: MFCD02094029