L19045.03
L19045.03EA
93
EUR
InStock
L19045.03
(R)-(+)-1-Octyne-3-ol ≥98%
(R)-(+)-1-Octyne-3-ol
Purity: 98+%
CAS No.: 32556-70-0
Molecular Formula: C8H14O
Formula Weight: 126.20
Flash Point: 63°(145°F)
Refractive Index: 1.4420
Boiling Point: 100°/20mm
Beilstein Reference: 2037704
Optical Rotation: +9.9° (neat)
MDL No.: MFCD00192370