L19068.03
L19068.MD
L19068.03EA
84.9
EUR
InStock
L19068.03
L19068.MD
Harmine ≥98%
Harmine
Purity: 98+%
CAS No.: 442-51-3
Molecular Formula: C13H12N2O
Formula Weight: 212.25
Odour: Odourless
Melting Point: 262-266°
Beilstein Reference: 178813
MDL No.: MFCD00004958