Synonyms:
Di(μ-chloro)dichlorobis(p-cymene)diruthenium(II)
L19126.04
L19126.09
L19126.ME
L19126.04EA
124
EUR
InStock
L19126.04
L19126.09
L19126.ME
Dichloro(p-cymene)ruthenium(II) dimer 98%
Dichloro(p-cymene)ruthenium(II) dimer
Purity: 98%
CAS No.: 52462-29-0
Molecular Formula: C20H28Cl4Ru2
Formula Weight: 612.40
MDL No.: MFCD00064793