L19249.03
L19249.03EA
59.1
EUR
InStock
L19249.03
Benzhydryl isothiocyanate ≥97%
Benzhydryl isothiocyanate
Purity: 97%
CAS No.: 3550-21-8
Molecular Formula: C14H11NS
Formula Weight: 225.31
Melting Point: 58-60°
Boiling Point: 160-165°/4mm
UN No.: 2811
MDL No.: MFCD00046815