L19310.14
L19310.14EA
511
EUR
InStock
L19310.14
(R)-(-)-2-Aminononane ≥99%, ee 98%, ChiPros®
(R)-(-)-2-Aminononane
Purity: 99+%
CAS No.: 74069-74-2
Molecular Formula: C9H21N
Formula Weight: 143.27
Flash Point: 71°(159°F)
Refractive Index: 1.4271
Boiling Point: 73°/19mm
Optical Rotation: -6° (neat)
UN No.: 2735
MDL No.: MFCD03844739