Synonyms:
3,5-Difluorobenzene-1-sulphonyl chloride
L19507.06
L19507.06EA
92.1
EUR
InStock
L19507.06
Chlorure de 3,5-difluorobenzènesulfonyle ≥97%
Chlorure de 3,5-difluorobenzènesulfonyle
Purity: 97%
CAS No.: 210532-25-5
Molecular Formula: C6H3ClF2O2S
Formula Weight: 212.60
Flash Point: 110°(230°F)
Melting Point: 57-59°
Beilstein Reference: 8053561
UN No.: 3261
MDL No.: MFCD02091378