L19554.03
L19554.03EA
91.8
EUR
InStock
L19554.03
2-(Trifluorométhyl)pyridine ≥99%
2-(Trifluorométhyl)pyridine
Purity: 99%
CAS No.: 368-48-9
Molecular Formula: C6H4F3N
Formula Weight: 147.10
Flash Point: 42°(108°F)
Refractive Index: 1.4180
Boiling Point: 139-141°
Beilstein Reference: 3112
UN No.: 1993
MDL No.: MFCD02183562