Synonyms:
2-Fluoro-6-hydroxybenzaldehyde
L19900.03
L19900.03EA
134
EUR
InStock
L19900.03
6-Fluorosalicylaldéhyde ≥97%
6-Fluorosalicylaldéhyde
Purity: 97%
CAS No.: 38226-10-7
Molecular Formula: C7H5FO2
Formula Weight: 140.12
Melting Point: 37-39°
Beilstein Reference: 1935289