L20240.03
L20240.03EA
76.2
EUR
InStock
L20240.03
(S)-(-)-1-(4-Methoxyphenyl)ethyl isocyanate 95%
(S)-(-)-1-(4-Methoxyphenyl)ethyl isocyanate
Purity: 95%
CAS No.: 745783-83-9
Molecular Formula: C10H11NO2
Formula Weight: 177.20
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5210
UN No.: 2206
MDL No.: MFCD05664085