L20245.03
L20245.03EA
134
EUR
InStock
L20245.03
(S)-(+)-2-Octyl isocyanate 95%
(S)-(+)-2-Octyl isocyanate
Purity: 95%
CAS No.: 745783-87-3
Molecular Formula: C9H17NO
Formula Weight: 155.24
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4230
Boiling Point: 68°/5mm
UN No.: 2206
MDL No.: MFCD05664089