Logic® Vue Class II Enclosures, for Protection and Containment for Large Instruments
Fournisseur: LABCONCO
3300411
LBCP3300411EA 0 EUR InStock
LBCP3300411
Logic® Vue Class II Enclosures, for Protection and Containment for Large Instruments
Hottes et coffrets Postes de sécurité biologique
The Logic® Vue is a specialised class II enclosure providing personnel and product protection for large automated instruments such as liquid handlers. The Logic® Vue provides laminar ISO 5 air.
  • Large capacity enclosure
  • Accommodates diverse range of instruments such as liquid handlers and cell sorters
  • Provides accessibility from front and back
  • Visibility from all sides
  • Powered by MyLogic™ OS to interface with a variety of devices for additional researcher safety
Certifications: Meets NSF/ANSI standard 49.
Conditionnement: The international packaging indicated for selected units includes reduced height for high cube shipping containers.
Attention: Installation required.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité