Ductless work stations, Protector®
Fournisseur: LABCONCO
3930020 3930021
LBCP3930020EA 0 EUR InStock
LBCP3930020 LBCP3930021
Ductless work stations, Protector®
Hottes et coffrets Hottes filtrantes
Protector® work stations are specifically designed for the needs of pathologists and histotechnologists. Like a traditional fume hood, these enclosures have a front air foil and rear baffle to direct the inflow air across the work area and away from the operator. Ideal for cramped or crowded labs, protector work stations have a low profile so they fit easily under standard upper cabinets. Designed with a built-in blower which use specially treated carbon filters.
  • Face velocities from 60 to 100 fpm
  • Built-in dynamically-balanced motorized impeller
  • Ergonomic
More Product Information
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité