Purifier® Vertical Clean Benches
Fournisseur: LABCONCO
3970424 3970220 3970224
LBCP3970424EA 0 EUR InStock
LBCP3970424 LBCP3970220 LBCP3970224
Purifier® Vertical Clean Benches
Hottes et coffrets Hottes à flux laminaire
Clean benches provide product protection from environmental contaminants for applications requiring a particulate-free work area, including plant tissue culture, electronic part inspection, syringe filling, medical device assembly, media plate preparation, and PCR.
Base stands (sold separately) are made of durable 4,4 cm thick, tubular, glacier white, epoxy-coated steel. The height of base stands is fully adjustable from 69,9 to 85,1 cm in 2,5 cm increments. Base stands with fixed feet come with levelling feet for use on uneven floor surfaces. Base stands with casters include four 12,7 cmMore Product Information
Commander maintenant
COMMANDE
SPÉCIFICATIONS
SERVICES
Services: VWRCATALYST™ offers customer on-site validation, testing & certification services including Face Velocity Profile Test, Smoke Generation Test, Tracer Gas Test, IQ, OQ, and PQ. Please call 888-793-2300 or email VWRCATALYST@vwr.com for more details. You can also visit https://vwr.com/equip_services.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité