Eyewash station
Fournisseur: LABORATOIRES ESCULAPE
8205203
LBES8205203EA 135 EUR InStock
LBES8205203
Eyewash station
Matériel de premiers secours Stations et douches pour le lavage oculaire
This eyewash station contains one 200 ml bottle with a pH neutral solution and one 500 ml bottle with an ophthalmic solution (sterile sodium chloride solution). The pH neutral solution is a sterile phosphate buffer (4,9%) that effectively and rapidly neutralises splashes of alkaline (base) or acidic solutions in the eyes or on the skin and which brings the pH level back to a normal level.
  • Neutral pH solution (blue bottle) for neutralising acids and alkalis
  • Sterile solution of sodium chloride (green bottle) for cleaning and rehydrating the eye
  • With mirror and pictograms
Rinsing time: 7 minutes
Shelf life: 3 years
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité