Pipette controller, FastPette™ V2
Fournisseur: Labnet International
P2000-230EU
LBNEP2000-230EUEA 0 EUR InStock
LBNEP2000-230EU
Pipette controller, FastPette™ V2
Pipettes Pipeteurs
This battery powered pipette controller works with glass and plastic pipettes in the 1 to 100 ml range.
  • Lightweight handle with smooth push-buttons and ergonomically located switches
  • High aspiration speed - 25 ml in 4 seconds
  • Cordless work for up to 8 continuous hours
  • Aspiration speed can be set to high or low to adjust for the volume of the pipette and viscosity of the liquid
  • Suction can be adjusted by the amount of pressure applied to the trigger button
  • Dispensing can be carried out by gravity or supported by the pump, which empties the pipette with a blow out step
  • Protected from the entrance of liquids by both a PTFE filter and a safety valve
  • Filter and pipette holder are easily exchanged and autoclaved
Information de livraison: Supplied with power supply/charger and PTFE filter.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité