Insect prepared slides
Fournisseur: LIEDER, JOHANNES
4300
LIED4300EA 0 EUR InStock
LIED4300
Insect prepared slides
Lames Lames préparées
Transverse section (t.s.).
  • Musca domestica, housefly, leaking-sucking mouth parts
  • Pieris, butterfly, sucking mouth parts
  • Carabus, ground beetle, biting mouth parts (carnivore)
  • Melolontha, cockchafer, chewing mouth parts (herbivore)
  • Pyrrhocoris, bug, piercing sucking mouth parts
  • Bombyx mori. silkworm moth, chewing mouth parts
  • Apis mellifica, honey bee, leaking sucking mouth parts of worker
  • Culex pipiens, mosquito, piercing sucking mouth parts
  • Melolontha, cockchafer, antenna with sense organs
  • Bombyx mori, silkworm moth, feathered antenna
  • Apis mellifica, anterior leg with eye brush
  • Apis mellifica, posterior leg with pollen basket
  • Musca domestica, house fly, leg with pulvilli
  • Apis mellifica, anterior and posterior wings
  • Pieris, butterfly, portion of wings with scales
  • Trachea from insect
  • Spiracle from insect
  • Cornea isolated from insect eye
  • Apis mellifica, honey bee, sting and poison sac
  • Apis mellifica, head with compound eyes and brain (t.s)
  • Bombyx mori, silkworm (t.s) showing silk spinning glands
  • Drosophila, fruit fly of adult
  • Ctenocephalus canis, dog flea of an adult
  • Culex pipiens, mosquito, larva
  • Chironomus, gnat, larva
Informations de commande: Includes 25 microscopic slides with accompanying brochure.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité