ACK Lysing Buffer (1x), TheraPEAK™
Fournisseur: LONZA
BP10-548E
LONZBP10-548EEA 37 EUR InStock
LONZBP10-548E
ACK Lysing Buffer (1x), TheraPEAK™
Buffers Tampons d'extraction
Ammonium-Chloride-Potassium (ACK) Lysing Buffer is used to lyse red blood cells in preparations containing white blood cells.
  • Quick and easy lysis of red blood cell
  • Isolation of white blood cells
All TheraPEAK™ Products are produced according to applicable GMP standards and follow the USP/EP guidance for cell and gene therapy raw materials. It is the end user's responsibility to ensure full compliance with local regulations in terms of safety assessments and effective removal prior to patient exposure.
TheraPEAKTM Media Products are produced according to applicable GMP raw material standards and are intended to support GMP manufacturing. TheraPEAKTM Media Products are produced at FDA registered manufacturing sites with an ISO 13485 certified quality management system.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité