Photometer, MD 600 with reagents for waste water
Fournisseur: Lovibond
214100
LOVI214100EA 3190 EUR InStock
LOVI214100
Photometer, MD 600 with reagents for waste water
Kits de tests environnementaux
MD 600 photometer is for precise and accurate measurements with a small footprint.
  • Complete and ready to use system
  • Ideal for site laboratories where accuracy, simplicity and space are key requirements
The setup uses Lovibond ® Vario tube tests and powder packs which enable the user to easily perform highly sensitive and precise water testing. It tests COD LR(0 to 150 mg/L), COD MR(0 to 1500 mg/L), Ammonia HR(1 to 50 mg/L N), Nitrate (1 to 30 mg/L), Nitrite (0,01 to 0,3 mg/L N), Nitrogen (Total 5 to 150 mg/L N) and Phosphate (0,02 to 1 mg/L P).
Information de livraison: It is supplied with MD 600 photometer and its accessories, RD 125 thermoreactor, IRiM infra-red module, tube, vial stand, membrane filter set, instruction manual and guarantee sheet.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité