Test kits, water balance, Comparator 2000+
Fournisseur: Lovibond
411290
LOVI411290EA 979 EUR InStock
LOVI411290
Test kits, water balance, Comparator 2000+
Kits de tests environnementaux
Comparator 2000+ is a versatile modular system for testing water.
  • Stable and accurate
  • Robust, compact and portable
  • Prism enabling colour matching
  • Diffuser plate is used to measure the turbidity
  • Longer path length cells be used for lower concentration analysis
The integrated prism brings the glass standards of the test discs and the coloured sample into the same field of view.
CHECKIT® disc contains a continuous colour scale which makes it possible to achieve an exact colour match between the colour standard and the sample. These discs are specially manufactured in selected materials to remain colour-stability over a long period and guarantee reliable, reproducible measurement results.
Information de livraison: Kit includes Comparator 2000+, pH comparator disc, chlorine comparator disc, 4× Comparator vials (10 ml), 13 mm path length shaker tubes, tablet reagents for each parmeter, crushing / stirring rod, vial cleaning brush, 100 ml graduated cylinder, instruction sheet and Langelier index / balanced water calculation table.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité