Test kit, AF129CYS, balanced water including cyanuric acid, Comparator 2000+
Fournisseur: Lovibond
411296
LOVI411296EA 964 EUR InStock
LOVI411296
Test kit, AF129CYS, balanced water including cyanuric acid, Comparator 2000+
Kits de tests environnementaux
Comparator 2000+ is a versatile modular system for testing water.This kit measures chlorine, alkalinity, pH calcium hardness and cyanuric acid.
  • Stable and accurate
  • Robust, compact and portable
  • Prism enabling colour matching
  • Diffuser plate is used to measure the turbidity
  • Longer path length cells be used for lower concentration analysis
Information de livraison: Test kit is supplied in a robust carrying case containing Comparator 2000+, pH comparator disc, chlorine comparator disc, 4× comparator vials, 10 ml, 13 mm path length, shaker tube, cyanuric acid tube, tablet reagents for each parmeter, crushing/stirring rod, vial cleaning brush, 100 ml graduated cylinder, instruction sheet, langelier index/balanced water and calculation table.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité