Drop test titration kits and reagent packs, MiniKit
Fournisseur: Lovibond
56K005501
LOVI56K005501EA 69.4 EUR InStock
LOVI56K005501
Drop test titration kits and reagent packs, MiniKit
Kits de tests environnementaux
Derived from laboratory burette methods, the drop test range is an established methodology used across industry and which forms an integral part of the analytical program of many a speciality chemical supplier. The reagents are quick to use and give results fast, making the drop test the method of choice for many customers.
Information de livraison: Kit is supplied with all reagents and equipment to perform the test, housed in a plastic case. Reagent pack is supplied in a carton containing test reagents and instruction sheet only.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité