Conductivity meter, handheld, SensoDirect Con110
Fournisseur: Lovibond
722300
LOVI722300EA 203 EUR InStock
LOVI722300
Conductivity meter, handheld, SensoDirect Con110
Electrochemistry Meters Conductimètres
The SensoDirect Con 110 is a simple, easy-to-use, portable meter for conductivity measurement.
  • Compact and versatile
  • With flip out stand for bench top use
  • Equipped as standard with a protective casing and built-in electrode holder
  • LC display shows two or three decimal places and a measuring range either of 0,001 to 1,999 or 0,01 to 19,99 mS/cm
  • Automatic temperature compensation feature.
  • Can be calibrated and adjusted using a potentiometer
Typical applications include: Drinking water, cooling/boiler water, waste water, pool water, surface water, water treatment companies, laboratories.
Information de livraison: Supplied with conductivity sensor, sturdy plastic case, battery, guarantee sheet and instruction manual.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
SERVICES
Services: Besoin d'une installation, d'un IQOQ, d'un entretien ou d'une réparation? Contactez-nous : technical.service.be@vwr.com
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité