M144-1LEA
104
EUR
InStock
M144-1L
TRITON™ X-114, grade dédié à la protéomique
Triton® X-114
Détergent non ionique qui sépare efficacement les protéines hydrophiles des protéines transmembranaires et hydrophobes sans affecter l'activité biologique.
- High purity and protease-free
- Suitable for most biological applications
These detergents have uncharged hydrophilic groups and are gentle, non-denaturing detergents ideal for studying the conformation and function of proteins, because they keep secondary and tertiary structure intact.