M147-1LEA
103
EUR
InStock
M147-1L
Tween® 20 (Polysorbate), grade dédié à la protéomique
Tween® 20 (Polysorbate)
Détergent non ionique qui sépare efficacement les protéines hydrophiles des protéines transmembranaires et hydrophobes sans affecter l'activité biologique.
- High purity and protease-free
- Suitable for most biological applications
Non-ionic detergents have uncharged hydrophilic groups and are gentle, non-denaturing detergents ideal for studying the conformation and function of proteins, because they keep secondary and tertiary structure intact.