M228-10ML-5PKEA
163
EUR
InStock
M228-10ML-5PK
Tween® 20 (Polysorbate) 10% en solution aqueuse, grade dédié à la protéomique exempt de peroxydes
Tween® 20 (Polysorbate)
Contains 5×10 ml vials of 10% solution.
- Optimised for isolating proteins and sample preparation
- Contains < 1ug of peroxide per ml
- Convenient concentrations pre-measured in ampoules
- Long-term stability
Information de livraison: Ce produit contient 5 flacons de 10 ml de solution 10%.