ReproSil XR C18 HPLC Column
Fournisseur: DR MAISCH
RX15.9E.S2503
MAISRX15.9E.S2503EA 290 EUR InStock
MAISRX15.9E.S2503
ReproSil XR C18 HPLC Column
Colonnes pour chromatographie
An all round and fully-porous, scalable spherical Silica from 1,5 to 15 µm with a high purity.
The metal content is less than 100 ppb. The silica impresses with its very narrow pore size distribution this make 99% of the surface accessible for the separation. The uniform bonded phase coverage translates to symmetrical peaks for acids/bases, and predictable reversed phase selectivity. Because of its very low metal content it also impressed with a high pH stability.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité