CHROMABOND® WAX SPE Columns
Fournisseur: MACHEREY-NAGEL
7300014
MANA7300014EA 120 EUR InStock
MANA7300014
CHROMABOND® WAX SPE Columns
Colonnes pour chromatographie
State-of-the-art spherical polymer for highest demands in solid phase extraction. PS/DVB copolymer with secondary and tertiary ammonium modification for mixed-mode SPE with weak anion exchange and reversed-phase properties (WAX).
High-purity material with ideal flow properties and highest reproducibility. Allows highest reproducibility with extremely low blind values due to an optimised production process.
Outstanding recovery rates especially for the enrichment of acidic analytes like short-chain PFAS.
Acidic analytes from various samples. Strong acids with pka < 1. Per- and polyfluoroalkyl substances (PFAS) from drinking water, soil, sediment, and wastewater.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité