NucleoBond Xtra BAC Buffer Set buffer set for the isolation of large constructs, like BACs, PACs, and P1, only applicable with Nucle oBond Xtra BAC kits - Buffer RES-BAC, Buffer LYS-BAC, Buffer NEU-BAC (1000 mL each), RNase A no dangerous goods acco 1 * 1 Pce
Fournisseur: MACHEREY-NAGEL
NucleoBond Xtra BAC Buffer Set buffer set for the isolation of large constructs, like BACs, PACs, and P1, only applicable with Nucle oBond Xtra BAC kits - Buffer RES-BAC, Buffer LYS-BAC, Buffer NEU-BAC (1000 mL each), RNase A no dangerous goods acco 1 * 1 Pce
COMMANDE
Développer tout / Réduire tout
NucleoBond Xtra BAC Buffer Set buffer set for the isolation of large constructs, like BACs, PACs, and P1, only applicable with Nucle oBond Xtra BAC kits - Buffer RES-BAC, Buffer LYS-BAC, Buffer NEU-BAC (1000 mL each), RNase A no dangerous goods acco 1 * 1 Pce MANA740437 1 * 1 Pce
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou