HPLC columns, PFAS Delay, NUCLEODUR®
Fournisseur: MACHEREY-NAGEL
760673.20
MANA760673.20EA 570 EUR InStock
MANA760673.20
HPLC columns, PFAS Delay, NUCLEODUR®
Colonnes pour chromatographie
NUCLEODUR PFAS Delay columns trap and therefor delay system-related PFAS contaminants, ensuring trace level analysis of PFAS in your samples. NUCLEODUR PFAS Delay reversed phase is hydrophobic with distinct polar selectivity for special PFAS analysis. Suitability for PFAS analysis is assured through qualified batch analysis.
  • High purity spherical silica gel
  • Polar selectivity
  • Special PFAS analysis
  • Low bleeding characteristics
This HPLC phase is made of high purity NUCLEODUR HPLC silica gel and is extensively endcapped. Due to a low bleeding characteristics these columns are suitable for high sensitivity LC/MS analysis.
Information de livraison: Supplied with a test chromatogram, instruction leaflet and batch test certificate.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité