Test paper, qualitative, Plumbtesmo®
Fournisseur: MACHEREY-NAGEL
90602
MANA90602EA 64.8 EUR InStock
MANA90602
Test paper, qualitative, Plumbtesmo®
Kits de tests environnementaux
The test paper allows the quick and easy detection of lead (Pb) on surfaces including metal surfaces, dishes, ceramics and toys. In criminal investigations it is used to search for traces of bullets.
  • Colour change: White-yellow to pink
The surface in question is moistened with distilled water. The test paper is pressed against the surface for about two minutes. In the presence of Lead the paper turns pink to dark purple.
Limit of sensitivity: 5 mg/L
Conditionnement: Pack of 40 sheets 40×25 mm
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité