Qualitative test paper peroxtesmo KM for blood traces (peroxidase)
Fournisseur: MACHEREY-NAGEL
90605
MANA90605EA 150 EUR InStock
MANA90605
Qualitative test paper peroxtesmo KM for blood traces (peroxidase)
Kits de tests environnementaux
This test paper allows the quick and sensitive detection of blood traces. In criminal investigations it is used as a rapid screening test.
  • Colour change: White to blue
The test paper uses the pseudo-peroxidatic effect of blood. The material in question is allowed to soak in water for about one minute.
Then the test paper is pressed to the sample. Blood traces lead to a distinct discolouration within a few seconds. The test papers are single sealed in plastic. This ensures safe results at any time.
Conditionnement: Pack of 25 test papers 15×30 mm.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité