Qualitative test paper peroxtesmo MI for peroxidase in milk
Fournisseur: MACHEREY-NAGEL
90627
MANA90627EA 30.1 EUR InStock
MANA90627
Qualitative test paper peroxtesmo MI for peroxidase in milk
Kits de tests environnementaux
This test paper allows the specific detection of lactoperoxidase in milk. In the dairy industry the test is used for the quick and easy quality control of highly heated milk. In contrast to the alternative guaiacol method, Peroxtesmo MI is odourless and non-toxic.
  • Colour change: White to blue
A drop of milk is put onto the test paper. If the paper remains white, the high heating process is completed.
Limit of sensitivity: 3% raw milk in UHT-milk
Conditionnement: Pack of 100 test papers 15×15 mm.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité