Test strips, cyanuric acid, swimming pool test
Fournisseur: MACHEREY-NAGEL
90710
MANA90710EA 13 EUR InStock
MANA90710
Test strips, cyanuric acid, swimming pool test
Kits de tests environnementaux
These test strips allow the quick and easy determination of the water quality in swimming pools. The test provides valuable results about the Cyanuric acid content.
  • Reliable result is obtained within 30 seconds.
  • Just dip, wait and read against multi-stage color scale
  • Always ready to use
  • No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
The test strip is dipped into the pool and moved back and forth 5 times.
Monitoring of the water quality allows adjusting the dosage of water conditioners in good time. The pool remains nice and clean.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité