Test strips, Ag-Fix
Fournisseur: MACHEREY-NAGEL
90741
MANA90741EA 32.2 EUR InStock
MANA90741
Test strips, Ag-Fix
Kits de tests environnementaux
With these test strips two paper zones are sealed onto the lower end of a plastic strip, one for the determination of the silver concentration and one for pH measurement. After dipping the test strip in the test solution the colours of the test zones are compared with the colour scale on the tube.
  • Always ready to use: no accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
  • The length of the strips has been chosen for safe and convenient handling
During operation of a fixing bath it is necessary to check the silver content as well as the pH in regular intervals to insure proper operation. A decrease in fixing capacity is noted as an increase in Silver concentration or an alteration of the pH value.
These strips allow a simultaneous semi quantitative determination of silver and the pH value in photographic solutions.
Colour reaction: Ag+: Ochreous to brown, pH : Yellow to blue
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité