Test strips, potassium iodide starch paper
Fournisseur: MACHEREY-NAGEL
90758
MANA90758EA 25.4 EUR InStock
MANA90758
Test strips, potassium iodide starch paper
Kits de tests environnementaux
This test paper allows the quick and easy detection of strong oxidisers such as Nitrite and free Chlorine. Potassium iodide starch paper is also used to control diazotisation reactions.
  • Nitrite: White to blue-violet
  • Chlorine: white to blue-violet
Nitrite or free chlorine oxidise potassium iodide to form elemental Iodine which reacts with starch to a blue-violet complex.
Potassium iodide starch paper may be used by dipping it into the sample or by applying drops of the sample onto the paper. To detect gaseous oxidisers, the paper is moistened with distilled water and then exposed to the gas in question.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité