Test paper, qualitative, sulphide
Fournisseur: MACHEREY-NAGEL
90761
MANA90761EA 14.7 EUR InStock
MANA90761
Test paper, qualitative, sulphide
Kits de tests environnementaux
The test paper allows the easy and reliable detection of hydrogen sulphide. This gas occurs in the processing of raw oil. It is toxic even in low concentrations. Therefore, critical points are carefully checked.
  • Colour change: White to brown-black
Sulphide ions in solution also give a positive reaction. The frequently used lead acetate paper is toxic and needs to be declared as hazardous. Sulphide test paper provides an alternative which is non toxic and safe.
Limit of sensitivity: 1 drop a solution containing 5mg/L sulphide (S²⁻) shows a positive reaction.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité