Arsenic test papers, qualitative
Fournisseur: MACHEREY-NAGEL
90762
MANA90762EA 43.6 EUR InStock
MANA90762
Arsenic test papers, qualitative
Kits de tests environnementaux
This test paper allows the easy detection of arsine (AsH₃) in the gas phase. Arsenic in solutions has to be converted into AsH₃ with Zn/acid and purged from the solution. The Arsine is detected directly at the boundary layer between water and air.
  • Colour change: White to brown-black
Arsenic test paper is used for the easy determination of arsenic in grape must and wine. For the determination of Arsine in compartment air the test paper is moistened with acetic anhydride.
Limit of sensitivity: 0,5 µg As
Information de livraison: Pack of 200 strips 20×70 mm.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité