Test strips, carbonate hardness, QUANTOFIX®
Fournisseur: MACHEREY-NAGEL
91323
MANA91323EA 32.6 EUR InStock
MANA91323
Test strips, carbonate hardness, QUANTOFIX®
Kits de tests environnementaux
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. The test strips are intended for single use, maintenance or calibration are not required.
  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
The test strips are used for the rapid and reliable determination of alkalinity in water. The easy dip-and-read-procedure provides a reliable result within 30 seconds. Carbonate hardness or alkalinity is a measure for the waters pH-buffer capacity. If the carbonate hardness is higher, addition of acids or bases will have a lower influence on the resulting pH. Sudden and rapid pH-changes of the water are avoided. QUANTOFIX® Carbonate hardness is used for the quick and easy control of water in swimming pools and aquariums.
Gradation: 0 · 3 · 6 · 10 · 15 · 20 °d
1 °d = 1,8 °f = 1,25 °e = 17,8 mg/l CaCO₃
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité